missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperidine-4-carbonitrile, 97%, Thermo Scientific™

Product Code. 10773843
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 25503-91-7
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.20
MDL Number MFCD00221055
InChI Key NFDGRMQIOHRQHF-UHFFFAOYSA-N
Synonym 1-acetyl-4-piperidinecarbonitrile, 1-acetyl-4-cyanopiperidine, 1-acetyl-isonipecotonitrile, 4-piperidinecarbonitrile,1-acetyl, 1-ethanoylpiperidine-4-carbonitrile
PubChem CID 2756286
IUPAC Name 1-acetylpiperidine-4-carbonitrile
SMILES CC(=O)N1CCC(CC1)C#N

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 152.2
Chemical Name or Material 1-Acetylpiperidine-4-carbonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.