missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanamine, 98%, Thermo Scientific Chemicals

£46.03 - £407.03

Chemical Identifiers

CAS 768-94-5
Molecular Formula C10H17N
Molecular Weight (g/mol) 151.253
MDL Number MFCD00074732
InChI Key DKNWSYNQZKUICI-UHFFFAOYSA-N
Synonym amantadine, 1-adamantanamine, 1-adamantylamine, 1-aminoadamantane, adamantanamine, adamantylamine, aminoadamantane, amantidine, symadine, symmetrel
PubChem CID 2130
ChEBI CHEBI:2618
IUPAC Name adamantan-1-amine
SMILES C1C2CC3CC1CC(C2)(C3)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15403867
View Documents
Thermo Scientific Alfa Aesar
H30076.06
5 g
£46.03
5g
Please to purchase this item. Need a web account? Register with us today!
15413867
View Documents
Thermo Scientific Alfa Aesar
H30076.14
25 g
£140.70
25g
Please to purchase this item. Need a web account? Register with us today!
15423867
View Documents
Thermo Scientific Alfa Aesar
H30076.22
100 g
£407.03
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Use only in a chemical fume hood. Keep away from sources of ignition. Incompatible with oxidizing agent.
Specifications

Chemical Identifiers

768-94-5
151.253
DKNWSYNQZKUICI-UHFFFAOYSA-N
2130
adamantan-1-amine
C10H17N
MFCD00074732
amantadine, 1-adamantanamine, 1-adamantylamine, 1-aminoadamantane, adamantanamine, adamantylamine, aminoadamantane, amantidine, symadine, symmetrel
CHEBI:2618
C1C2CC3CC1CC(C2)(C3)N

Specifications

768-94-5
C10H17N
amantadine, 1-adamantanamine, 1-adamantylamine, 1-aminoadamantane, adamantanamine, adamantylamine, aminoadamantane, amantidine, symadine, symmetrel
DKNWSYNQZKUICI-UHFFFAOYSA-N
adamantan-1-amine
2130
151.25
1-Adamantanamine
∼200°C (sublimation)
MFCD00074732
Soluble in organic solvents. Insoluble in water.
C1C2CC3CC1CC(C2)(C3)N
151.253
CHEBI:2618
98%
Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, Causes skin irritation

Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)

missing translation for 'einecsNumber' : 212-201-2

missing translation for 'rtecsNumber' : YD1925000

missing translation for 'tsca' : Yes

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only