missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanemethylamine, 98%, Thermo Scientific Chemicals

£79.90 - £267.33

Chemical Identifiers

CAS 17768-41-1
Molecular Formula C11H20N
Molecular Weight (g/mol) 166.29
MDL Number MFCD00074750
InChI Key XSOHXMFFSKTSIT-UHFFFAOYSA-O
Synonym 1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine
PubChem CID 86625
IUPAC Name 1-adamantylmethanamine
SMILES [NH3+]CC12CC3CC(CC(C3)C1)C2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10695612
View Documents
Thermo Scientific Acros
177420010
1 g Glass bottle
£79.90
1g
Please to purchase this item. Need a web account? Register with us today!
10219163
View Documents
Thermo Scientific Acros
177420050
5 g Glass bottle
£267.33
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17768-41-1
166.29
XSOHXMFFSKTSIT-UHFFFAOYSA-O
86625
[NH3+]CC12CC3CC(CC(C3)C1)C2
C11H20N
MFCD00074750
1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine
1-adamantylmethanamine

Specifications

17768-41-1
100.0
0.9300g/mL
92°C
98%
1.5127 to 1.5147
0.93
XSOHXMFFSKTSIT-UHFFFAOYSA-O
1-adamantylmethanamine
86625
98%
1-Adamantanemethylamine
97.5
Colorless to Yellow
83.0°C to 85.0°C (0.3 mmHg)
Authentic
C11H20N
MFCD00074750
1-adamantanemethylamine, adamantan-1-ylmethanamine, 1-aminomethyl adamantane, 1-aminomethyladamantane, 1-aminomethyl-adamantane, 1-1-adamantyl methanamine, 1-adamantan-1-ylmethanamine, c-adamantan-1-yl-methylamine, 1-adamantanemethyl amine, tricyclo 3.3.1.13,7 dec-1-ylmethylamine
[NH3+]CC12CC3CC(CC(C3)C1)C2
166.29
165.28
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 241-752-1

Product Identifier
  • 1-Adamantanemethylamine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only