missing translation for 'onlineSavingsMsg'
Learn More

1-Allylpiperidine, 97%, Thermo Scientific™

Product Code. 15408959
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15408959

Brand: Thermo Scientific Alfa Aesar L12312.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Allylpiperidine used to produce 4-chloro-2-piperidin-1-yl-quinazoline at the temperature of 100°C.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. St
TRUSTED_SUSTAINABILITY

Specifications

CAS 14446-67-4
Boiling Point 151°C to 152°C
Molecular Formula C8H15N
Refractive Index 1.458
MDL Number MFCD00078381
Quantity 5 g
UN Number UN1993
Beilstein 107480
Synonym 1-allylpiperidine, n-allylpiperidine, 1-2-propenyl piperidine, n-allyl piperidine, 1-prop-2-en-1-yl piperidine, n-allylpiperidin, 1-allyl-piperidine, acmc-1bt7q, piperidine, 1-2-propenyl
Solubility Information Not miscible or difficult to mix in water.
InChI Key KYGMSGYKSGNPHM-UHFFFAOYSA-N
SMILES C=CCN1CCCCC1
IUPAC Name 1-prop-2-enylpiperidine
Molecular Weight (g/mol) 125.215
PubChem CID 139756
Formula Weight 125.22
Percent Purity 97%
Chemical Name or Material 1-Allylpiperidine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.