missing translation for 'onlineSavingsMsg'
Learn More
1-Amino-1-cyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals Product Code.: 15417675

1-Amino-1-cyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 15417675
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15417675

Brand: Thermo Scientific Alfa Aesar H27264.MD

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68781-13-5
Molecular Formula C4H8ClNO2
Molecular Weight (g/mol) 137.563
MDL Number MFCD00012545
InChI Key WTZRHZRMLYNBRM-UHFFFAOYSA-N
Synonym 1-aminocyclopropanecarboxylic acid hydrochloride, 1-aminocyclopropane-1-carboxylic acid hydrochloride, 1-amino-cyclopropane-1-carboxylic acid hydrochloride, 1-aminocyclopropanecarboxylic acid hcl, cyclopropanecarboxylic acid, 1-amino-, hydrochloride, 1-aminocyclopropanecarboxylic acid, chloride, acpc-hcl, pubchem13872, ksc352q0j, 1-amino-cyclopropane-1-carboxylic acid, hcl
PubChem CID 11957445
IUPAC Name 1-aminocyclopropane-1-carboxylic acid;hydrochloride
SMILES C1CC1(C(=O)O)N.Cl

Specifications

Melting Point ∽220°C (decomposition)
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 137.56
Percent Purity 97%
Chemical Name or Material 1-Amino-1-cyclopropanecarboxylic acid hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Amino-1-cyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.