missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-1-cyclopropanecarboxylic acid hydrochloride, 97%, Thermo Scientific Chemicals

£116.00 - £321.00

Chemical Identifiers

CAS 68781-13-5
Molecular Formula C4H8ClNO2
Molecular Weight (g/mol) 137.563
MDL Number MFCD00012545
InChI Key WTZRHZRMLYNBRM-UHFFFAOYSA-N
Synonym 1-aminocyclopropanecarboxylic acid hydrochloride, 1-aminocyclopropane-1-carboxylic acid hydrochloride, 1-amino-cyclopropane-1-carboxylic acid hydrochloride, 1-aminocyclopropanecarboxylic acid hcl, cyclopropanecarboxylic acid, 1-amino-, hydrochloride, 1-aminocyclopropanecarboxylic acid, chloride, acpc-hcl, pubchem13872, ksc352q0j, 1-amino-cyclopropane-1-carboxylic acid, hcl
PubChem CID 11957445
IUPAC Name 1-aminocyclopropane-1-carboxylic acid;hydrochloride
SMILES C1CC1(C(=O)O)N.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15417675
View Documents
Thermo Scientific Alfa Aesar
H27264.MD
250 mg
£116.00
250mg
Please to purchase this item. Need a web account? Register with us today!
11312299
View Documents
Thermo Scientific Alfa Aesar
H27264.03
1 g
£321.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

68781-13-5
137.563
WTZRHZRMLYNBRM-UHFFFAOYSA-N
11957445
C1CC1(C(=O)O)N.Cl
C4H8ClNO2
MFCD00012545
1-aminocyclopropanecarboxylic acid hydrochloride, 1-aminocyclopropane-1-carboxylic acid hydrochloride, 1-amino-cyclopropane-1-carboxylic acid hydrochloride, 1-aminocyclopropanecarboxylic acid hcl, cyclopropanecarboxylic acid, 1-amino-, hydrochloride, 1-aminocyclopropanecarboxylic acid, chloride, acpc-hcl, pubchem13872, ksc352q0j, 1-amino-cyclopropane-1-carboxylic acid, hcl
1-aminocyclopropane-1-carboxylic acid;hydrochloride

Specifications

68781-13-5
C4H8ClNO2
Hygroscopic
WTZRHZRMLYNBRM-UHFFFAOYSA-N
1-aminocyclopropane-1-carboxylic acid;hydrochloride
11957445
97%
∽220°C (decomposition)
MFCD00012545
1-aminocyclopropanecarboxylic acid hydrochloride, 1-aminocyclopropane-1-carboxylic acid hydrochloride, 1-amino-cyclopropane-1-carboxylic acid hydrochloride, 1-aminocyclopropanecarboxylic acid hcl, cyclopropanecarboxylic acid, 1-amino-, hydrochloride, 1-aminocyclopropanecarboxylic acid, chloride, acpc-hcl, pubchem13872, ksc352q0j, 1-amino-cyclopropane-1-carboxylic acid, hcl
C1CC1(C(=O)O)N.Cl
137.563
137.56
1-Amino-1-cyclopropanecarboxylic acid hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only