missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aminopyrene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
Description
It acts as a precursor molecule for production of other fluorescent probes such as 1-Azidopyrene, APTS, and N-(1-Pyrene)iodoacetamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 116°C to 119°C |
Quantity | 0.5 g |
Solubility Information | Insoluble in water (0.576mg/L at 25 deg C). |
Formula Weight | 217.27 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | 1-Aminopyrene |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Signal Word
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'