missing translation for 'onlineSavingsMsg'
Learn More

1-Aza-15-crown-5, 97%, Thermo Scientific Chemicals

Product Code. 15457947
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Aza-15-crown-5 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofuctionalized macrocycles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aza-15-crown-5 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofuctionalized macrocycles. It is also used as pharmaceutical intermediate.

Solubility
Soluble in water and methanol.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66943-05-3
Molecular Formula C10H21NO4
Molecular Weight (g/mol) 219.281
MDL Number MFCD00075465
InChI Key BJUOQSZSDIHZNP-UHFFFAOYSA-N
Synonym aza-15-crown-5, 1-aza-15-crown 5-ether, 1-aza-15-crown-5, 1-aza-15-crown, 1-aza-4,7,10,13-tetraoxacyclopentadecane
PubChem CID 544820
IUPAC Name 1,4,7,10-tetraoxa-13-azacyclopentadecane
SMILES C1COCCOCCOCCOCCN1

Specifications

Quantity 250 mg
Beilstein 971959
Sensitivity Air Sensitive
Solubility Information Soluble in water and methanol.
Formula Weight 219.28
Percent Purity 97%
Chemical Name or Material 1-Aza-15-crown-5

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'