missing translation for 'onlineSavingsMsg'
Learn More

1-Benzofuran-2-sulfonyl chloride, Thermo Scientific™

Product Code. 10464312
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 17070-58-5
Molecular Formula C8H5ClO3S
Molecular Weight (g/mol) 216.635
InChI Key RWCKBBSBRTUUHR-UHFFFAOYSA-N
Synonym benzofuran-2-sulfonyl chloride, 2-benzofuransulfonylchloride, 2-benzofuransulfonyl chloride, benzo b furan-2-sulfonyl chloride, pubchem7024, benzo d furan-2-ylchlorosulfone
PubChem CID 2795172
IUPAC Name 1-benzofuran-2-sulfonyl chloride
SMILES C1=CC=C2C(=C1)C=C(O2)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 216.64
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-2-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'