missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10065972
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10065972

Brand: Thermo Scientific Maybridge CC24804CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10133-30-9
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD05663673
InChI Key QHHRWAPVYHRAJA-UHFFFAOYSA-N
Synonym benzo b thiophene-5-carbaldehyde, benzo b thiophene-5-carboxaldehyde, 5-formylbenzo b thiophene, 1-benzothiophene-5-carboxaldehyde, benzothiophene-5-carbaldehyde, 5-thionaphthenecarbaldehyde
PubChem CID 139097
IUPAC Name 1-benzothiophene-5-carbaldehyde
SMILES O=CC1=CC=C2SC=CC2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 162.21
Percent Purity 95%
Chemical Name or Material 1-Benzothiophene-5-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.