missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-7-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10397064
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10134-98-2
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.205
InChI Key LJPSRTWIAXVPIS-UHFFFAOYSA-N
Synonym benzo b thiophene-7-carboxylic acid, benzo b thiophene-7-carboxylicacid, 7-carboxybenzo b thiophene, 1-benzo b thiophene-7-carboxylic acid
PubChem CID 14235073
IUPAC Name 1-benzothiophene-7-carboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)SC=C2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-7-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'