missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoyl-4-piperidone, 98+%, Thermo Scientific Chemicals

£44.70 - £388.00

Chemical Identifiers

CAS 24686-78-0
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.24
MDL Number MFCD00006189
InChI Key NZAXGZYPZGEVBD-UHFFFAOYSA-N
Synonym n-benzoyl-4-piperidone, 1-benzoyl-4-piperidinone, 4-piperidinone, 1-benzoyl, 1-benzoyl-4-piperidone, 1-benzoyl-4-oxopiperidine, 4-piperidone, 1-benzoyl, 1-phenylcarbonyl piperidin-4-one, 1-benzoyl-piperidin-4-one, n-benzoyl-4piperidone, 1-benzoyl-piperidinoine
PubChem CID 90583
IUPAC Name 1-benzoylpiperidin-4-one
SMILES O=C(N1CCC(=O)CC1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11461608
View Documents
Thermo Scientific Alfa Aesar
A13901.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11481608
View Documents
Thermo Scientific Alfa Aesar
A13901.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24686-78-0
203.24
NZAXGZYPZGEVBD-UHFFFAOYSA-N
90583
O=C(N1CCC(=O)CC1)C1=CC=CC=C1
C12H13NO2
MFCD00006189
n-benzoyl-4-piperidone, 1-benzoyl-4-piperidinone, 4-piperidinone, 1-benzoyl, 1-benzoyl-4-piperidone, 1-benzoyl-4-oxopiperidine, 4-piperidone, 1-benzoyl, 1-phenylcarbonyl piperidin-4-one, 1-benzoyl-piperidin-4-one, n-benzoyl-4piperidone, 1-benzoyl-piperidinoine
1-benzoylpiperidin-4-one

Specifications

24686-78-0
158°C to 160°C (0.2 mmHg)
C12H13NO2
158848
NZAXGZYPZGEVBD-UHFFFAOYSA-N
1-benzoylpiperidin-4-one
90583
≥98%
52°C to 57°C
>110°C (230°F)
MFCD00006189
n-benzoyl-4-piperidone, 1-benzoyl-4-piperidinone, 4-piperidinone, 1-benzoyl, 1-benzoyl-4-piperidone, 1-benzoyl-4-oxopiperidine, 4-piperidone, 1-benzoyl, 1-phenylcarbonyl piperidin-4-one, 1-benzoyl-piperidin-4-one, n-benzoyl-4piperidone, 1-benzoyl-piperidinoine
O=C(N1CCC(=O)CC1)C1=CC=CC=C1
203.24
203.24
1-Benzoyl-4-piperidone
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 246-407-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.