missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoylnaphthalene, 97%, Thermo Scientific™

Product Code. 11459637
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 642-29-5
Molecular Formula C17H12O
Molecular Weight (g/mol) 232.282
MDL Number MFCD00046431
InChI Key CXAYOCVHDCXPAI-UHFFFAOYSA-N
Synonym 1-benzoylnaphthalene, 1-naphthyl phenyl ketone, benzoylnaphthalene, naphthalen-1-yl phenyl methanone, naphthyl phenyl ketone, 1-naphthyl phenyl methanone, alpha-benzoylnaphthalene, methanone, 1-naphthalenylphenyl, naphthylphenylketone
PubChem CID 69503
IUPAC Name naphthalen-1-yl(phenyl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC3=CC=CC=C32

Specifications

Melting Point 75°C to 76°C
Quantity 5 g
Formula Weight 232.28
Percent Purity 97%
Chemical Name or Material 1-Benzoylnaphthalene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'