missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-carbethoxy-4-piperidone hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 10428630
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1454-53-1
Molecular Formula C15H20ClNO3
Molecular Weight (g/mol) 297.78
MDL Number MFCD00012800
InChI Key YPFMNHZRNXPYBG-UHFFFAOYNA-N
Synonym ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 1-benzyl-3-carbethoxy-4-piperidone hydrochloride, ethyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride, 1-benzyl-3-carbethoxy-4-piperidone hcl, 1-benzyl-3-carbethoxy-piperidone hcl, 1-benzyl-3-ethoxycarbonyl-4-piperidone hcl, 1-benzyl-3-carboethoxy-4-piperidone hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-oxopiperidine hydrochloride
PubChem CID 102623
IUPAC Name ethyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
SMILES Cl.CCOC(=O)C1CN(CC2=CC=CC=C2)CCC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 160.0°C to 170.0°C
Color Cream to Pink
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Quantity 25 g
Beilstein 22, II, 216
Formula Weight 297.77
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Benzyl-3-carbethoxy-4-piperidone hydrochloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Benzyl-3-carbethoxy-4-piperidone hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'