missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-pyrrolidinone, 98%, Thermo Scientific Chemicals

Product Code. 11424200
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775-16-6
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00005342
InChI Key DHGMDHQNUNRMIN-UHFFFAOYSA-N
Synonym 1-benzyl-3-pyrrolidinone, n-benzyl-3-pyrrolidinone, 1-phenylmethyl-3-pyrrolidinone, 1-benzyl-3-pyrrolidone, 1-benzyl-pyrrolidin-3-one, 1-benzyl pyrrolidin-3-one, 3-pyrrolidinone, 1-phenylmethyl, benzyl-3-pyrrolidinone, pubchem10007, n-benzylpyrrolid-3-one
PubChem CID 69890
IUPAC Name 1-benzylpyrrolidin-3-one
SMILES C1CN(CC1=O)CC2=CC=CC=C2

Specifications

Density 1.091
Boiling Point 77°C to 78°C (0.01 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 175.23
Percent Purity 98%
Chemical Name or Material 1-Benzyl-3-pyrrolidinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'