missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, tech. 90%, Thermo Scientific™

Product Code. 11331927
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52763-21-0
Molecular Formula C15H20ClNO3
Molecular Weight (g/mol) 297.78
MDL Number MFCD00012792
InChI Key UQOMEAWPKSISII-UHFFFAOYNA-N
Synonym ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, ethyl n-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride, ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 1-benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hcl, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hydrochloride, 1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride, ethyl n-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, zlchem 420, pubchem19671
PubChem CID 2723880
IUPAC Name ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)C1CCN(CC2=CC=CC=C2)CC1=O

Specifications

Melting Point ∽158°C (decomposition)
Quantity 5 g
Beilstein 3749159
Formula Weight 297.78
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.