missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Benzyl-4-piperidyl)methanol, Thermo Scientific™, Thermo Scientific™
£10.40 - £35.70
Chemical Identifiers
CAS | 67686-01-5 |
---|---|
Molecular Formula | C13H19NO |
Molecular Weight (g/mol) | 205.301 |
MDL Number | MFCD00831014 |
InChI Key | FLQPYEOKVZYXRL-UHFFFAOYSA-N |
Synonym | 1-benzyl-4-piperidyl methanol, 1-benzylpiperidin-4-yl methanol, 1-benzyl-4-piperidinemethanol, 1-benzyl-4-piperidinyl methanol, 1-benzyl-4-hydroxymethylpiperidine, 4-piperidinemethanol, 1-phenylmethyl, 1-benzy-4-hydroxymethylpiperidine, 1-benzy-4-hydroxymethyl piperidine, pubchem8894, 1-benzyl-4-piperdinemethanol |
PubChem CID | 736802 |
IUPAC Name | (1-benzylpiperidin-4-yl)methanol |
SMILES | C1CN(CCC1CO)CC2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10561132
|
Thermo Scientific Maybridge
KM09623DA |
1 g |
£10.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10061194
|
Thermo Scientific Maybridge
KM09623DE |
5 g |
£35.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
67686-01-5 | |
205.301 | |
FLQPYEOKVZYXRL-UHFFFAOYSA-N | |
736802 | |
C1CN(CCC1CO)CC2=CC=CC=C2 |
C13H19NO | |
MFCD00831014 | |
1-benzyl-4-piperidyl methanol, 1-benzylpiperidin-4-yl methanol, 1-benzyl-4-piperidinemethanol, 1-benzyl-4-piperidinyl methanol, 1-benzyl-4-hydroxymethylpiperidine, 4-piperidinemethanol, 1-phenylmethyl, 1-benzy-4-hydroxymethylpiperidine, 1-benzy-4-hydroxymethyl piperidine, pubchem8894, 1-benzyl-4-piperdinemethanol | |
(1-benzylpiperidin-4-yl)methanol |
Specifications
67686-01-5 | |
97% | |
C13H19NO | |
1-benzyl-4-piperidyl methanol, 1-benzylpiperidin-4-yl methanol, 1-benzyl-4-piperidinemethanol, 1-benzyl-4-piperidinyl methanol, 1-benzyl-4-hydroxymethylpiperidine, 4-piperidinemethanol, 1-phenylmethyl, 1-benzy-4-hydroxymethylpiperidine, 1-benzy-4-hydroxymethyl piperidine, pubchem8894, 1-benzyl-4-piperdinemethanol | |
C1CN(CCC1CO)CC2=CC=CC=C2 | |
205.301 | |
205.3 | |
(1-Benzyl-4-piperidyl)methanol |
97.0 | |
Amber glass bottle | |
MFCD00831014 | |
FLQPYEOKVZYXRL-UHFFFAOYSA-N | |
(1-benzylpiperidin-4-yl)methanol | |
736802 | |
97% |