missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyloxy-4-iodobenzene, 98+%, Thermo Scientific Chemicals
£52.60 - £215.00
Chemical Identifiers
CAS | 19578-68-8 |
---|---|
Molecular Formula | C13H11IO |
Molecular Weight (g/mol) | 310.134 |
MDL Number | MFCD00079704 |
InChI Key | MPWFGAWFTAZWKZ-UHFFFAOYSA-N |
PubChem CID | 519671 |
IUPAC Name | 1-iodo-4-phenylmethoxybenzene |
SMILES | C1=CC=C(C=C1)COC2=CC=C(C=C2)I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11332887
|
Thermo Scientific Alfa Aesar
L16092.06 |
5 g |
£52.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11342887
|
Thermo Scientific Alfa Aesar
L16092.14 |
25 g |
£215.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with oxidizing agents.
Chemical Identifiers
19578-68-8 | |
310.134 | |
MPWFGAWFTAZWKZ-UHFFFAOYSA-N | |
1-iodo-4-phenylmethoxybenzene |
C13H11IO | |
MFCD00079704 | |
519671 | |
C1=CC=C(C=C1)COC2=CC=C(C=C2)I |
Specifications
19578-68-8 | |
140°C (1 mmHg) | |
MFCD00079704 | |
Light sensitive | |
Insoluble in water. | |
C1=CC=C(C=C1)COC2=CC=C(C=C2)I | |
310.134 | |
310.14 | |
1-Benzyloxy-4-iodobenzene |
60°C to 64°C | |
C13H11IO | |
3051495 | |
1-benzyloxy-4-iodobenzene, 4-benzyloxyiodobenzene, 4-iodobenzyloxybenzene, benzyl 4-iodophenyl ether, benzene, 1-iodo-4-phenylmethoxy, 1-benzyloxy-4-iodo-benzene, benzene,1-iodo-4-phenylmethoxy, acmc-20amon, 4-benzyloxy iodobenzene | |
MPWFGAWFTAZWKZ-UHFFFAOYSA-N | |
1-iodo-4-phenylmethoxybenzene | |
519671 | |
≥98% |
Safety and Handling
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only