missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxycarbonyl-(2S,4R)-2-cyano-4-fluoropyrrolidine, 97%, Thermo Scientific™

Product Code. 15427705
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1301706-46-6
Molecular Formula C13H13FN2O2
MDL Number MFCD08458685
Synonym n-cbz-trans-4-fluoro-l-prolinonitrile, 1-benzyloxycarbonyl-2s,4r-2-cyano-4-fluoropyrrolidine, benzyl 2s,4r-2-cyano-4-fluoropyrrolidine-1-carboxylate, n-benzyloxycarbonyl-trans-4-fluoro-l-prolinonitrile, 2s,4r-benzyl 2-cyano-4-fluoropyrrolidine-1-carboxylate, benzyl 2s,4r-2-cyano-4-fluoro-pyrrolidine-1-carboxylate

Specifications

Melting Point 68°C to 72°C
Quantity 250 mg
UN Number UN3439
Formula Weight 248.25
Percent Purity 97%
Chemical Name or Material 1-Benzyloxycarbonyl-(2S,4R)-2-cyano-4-fluoropyrrolidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'