missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylpyrrole, 96%, Thermo Scientific Chemicals

Product Code. 15462338
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-97-0
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.22
MDL Number MFCD00963301
InChI Key FNEQHKCQXDKYEO-UHFFFAOYSA-N
Synonym n-benzylpyrrole, 1h-pyrrole, 1-phenylmethyl, 1-benzyl-1h-pyrrole, pyrrole, 1-benzyl, 1-benzylpyrrol, acmc-209fbl, 1-phenylmethyl pyrrole, 1-benzyl-1h-pyrrole #, pyrrole, 1-benzyl-8ci
PubChem CID 74922
SMILES C(N1C=CC=C1)C1=CC=CC=C1

Specifications

Color Colorless to Brown
Density 1.020 g/mL
Boiling Point 90°C (0.9 mmHg)
Refractive Index 1.5685
Quantity 1 g
UN Number UN1993
IUPAC Name 1-benzyl-1H-pyrrole
Formula Weight 157.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Benzylpyrrole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'