missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-1,2,3,6-tetrahydropyridine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Beschreibung
1-Boc-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Boc-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
1-Boc-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Spezifikation
Spezifikation
Boiling Point | 78°C to 84°C (5mmHg) |
Solubility Information | Insoluble in water. Soluble in organic solvents. |
Formula Weight | 183.25 |
Percent Purity | 97% |
Density | 1.029 |
Quantity | 250 mg |
Chemical Name or Material | 1-Boc-1,2,3,6-tetrahydropyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'