missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-fluoro-4-pyrrolidinone, Thermo Scientific Chemicals

Product Code. 11446150
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845894-03-3
Molecular Formula C9H14FNO3
Molecular Weight (g/mol) 203.21
MDL Number MFCD08274459
InChI Key HPHVNLXMQXEDFX-UHFFFAOYNA-N
Synonym 1-boc-3-fluoro-4-pyrrolidinone, boc-3-fluoro-4-oxopyrrolidine, 3-fluoro-4-oxopyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-4-fluoro-3-pyrrolidinone, n-t-boc-3-fluoro-4-oxopyrrolidine, tert-butyl3-fluoro-4-oxopyrrolidine-1-carboxylate
PubChem CID 45158916
IUPAC Name tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(F)C(=O)C1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 203.22
Chemical Name or Material 1-Boc-3-fluoro-4-pyrrolidinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'