missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-3-fluoro-4-pyrrolidinone, Thermo Scientific Chemicals
£239.00 - £660.00
Chemical Identifiers
CAS | 845894-03-3 |
---|---|
Molecular Formula | C9H14FNO3 |
Molecular Weight (g/mol) | 203.21 |
MDL Number | MFCD08274459 |
InChI Key | HPHVNLXMQXEDFX-UHFFFAOYNA-N |
Synonym | 1-boc-3-fluoro-4-pyrrolidinone, boc-3-fluoro-4-oxopyrrolidine, 3-fluoro-4-oxopyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-4-fluoro-3-pyrrolidinone, n-t-boc-3-fluoro-4-oxopyrrolidine, tert-butyl3-fluoro-4-oxopyrrolidine-1-carboxylate |
PubChem CID | 45158916 |
IUPAC Name | tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CC(F)C(=O)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15413607
|
Thermo Scientific Alfa Aesar
H27082.MD |
250 mg |
£239.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11446150
|
Thermo Scientific Alfa Aesar
H27082.03 |
1 g |
£660.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Boc-3-fluoro-4-pyrrolidinone is used as an intermediate in the manufacture of various substances such as pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
Chemical Identifiers
845894-03-3 | |
203.21 | |
HPHVNLXMQXEDFX-UHFFFAOYNA-N | |
45158916 | |
CC(C)(C)OC(=O)N1CC(F)C(=O)C1 |
C9H14FNO3 | |
MFCD08274459 | |
1-boc-3-fluoro-4-pyrrolidinone, boc-3-fluoro-4-oxopyrrolidine, 3-fluoro-4-oxopyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-4-fluoro-3-pyrrolidinone, n-t-boc-3-fluoro-4-oxopyrrolidine, tert-butyl3-fluoro-4-oxopyrrolidine-1-carboxylate | |
tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate |
Specifications
845894-03-3 | |
MFCD08274459 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)N1CC(F)C(=O)C1 | |
203.21 | |
203.22 |
C9H14FNO3 | |
1-boc-3-fluoro-4-pyrrolidinone, boc-3-fluoro-4-oxopyrrolidine, 3-fluoro-4-oxopyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-4-fluoro-3-pyrrolidinone, n-t-boc-3-fluoro-4-oxopyrrolidine, tert-butyl3-fluoro-4-oxopyrrolidine-1-carboxylate | |
HPHVNLXMQXEDFX-UHFFFAOYNA-N | |
tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate | |
45158916 | |
1-Boc-3-fluoro-4-pyrrolidinone |
Safety and Handling
missing translation for 'tsca' : No
RUO – Research Use Only