missing translation for 'onlineSavingsMsg'
Learn More
1-Boc-3-iodoazetidine, 97%, Thermo Scientific™ Product Code.: 15409307

1-Boc-3-iodoazetidine, 97%, Thermo Scientific™

Product Code. 15409307
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15409307

Brand: Thermo Scientific Alfa Aesar H52558.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Boc-3-iodoazetidine is used as an organic chemical synthesis intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 254454-54-1
Molecular Formula C8H14INO2
MDL Number MFCD09752821
Quantity 1 g
Sensitivity Light sensitive
Synonym 1-boc-3-iodoazetidine, 1-boc-3-iodo-azetidine, 1-boc-3-iodo azetidine, n-boc-3-iodoazetidine, 3-iodo-azetidine-1-carboxylic acid tert-butyl ester, 3-iodoazetidine, n-boc protected, 1-n-boc-3-iodoazetidine, 1-tert-butoxycarbonyl-3-iodoazetidine, tert-butyl 3-iodo-1-azetidinecarboxylate, tert-butyl 3-iodoazetidinecarboxylate
InChI Key XPDIKRMPZNLBAC-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CC(I)C1
IUPAC Name tert-butyl 3-iodoazetidine-1-carboxylate
Molecular Weight (g/mol) 283.11
PubChem CID 11000522
Formula Weight 283.11
Percent Purity 97%
Chemical Name or Material 1-Boc-3-iodoazetidine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Boc-3-iodoazetidine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.