missing translation for 'onlineSavingsMsg'
Learn More
1-Boc-3-(iodomethyl)azetidine, 95%, Thermo Scientific™ Product Code.: 15423968

1-Boc-3-(iodomethyl)azetidine, 95%, Thermo Scientific™

Product Code. 15423968
250 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 15423968

Brand: Thermo Scientific Alfa Aesar H57500.MD

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253176-94-2
Molecular Formula C9H16INO2
Molecular Weight (g/mol) 297.14
MDL Number MFCD09951816
InChI Key CJVGFEARJOREHI-UHFFFAOYSA-N
PubChem CID 53249943
IUPAC Name tert-butyl 3-(iodomethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(CI)C1

Specifications

Quantity 250 mg
Synonym tert-butyl 3-iodomethyl azetidine-1-carboxylate, 1-boc-3-iodomethyl azetidine, n-boc-3-iodomethyl azetidine, tert-butyl3-iodomethyl azetidine-1-carboxylate, 1-azetidinecarboxylic acid, 3-iodomethyl-, 1,1-dimethylethyl ester, acmc-209gj7, 1-n-boc-3-iodomethyl azetidine, tert-butyl-3-iodomethyl azetidin-1-carboxylate, tert-butyl 3-iodomethyl ;azetidine-1-carboxylate, tert-butyl 3-iodanylmethyl azetidine-1-carboxylate
Formula Weight 297.13
Percent Purity 95%
Chemical Name or Material 1-Boc-3-(iodomethyl)azetidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Boc-3-(iodomethyl)azetidine, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.