missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-formylphenyl)piperazine, 97%, Thermo Scientific™

Product Code. 15472368
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174855-57-3
Molecular Formula C16H22N2O3
Molecular Weight (g/mol) 290.363
MDL Number MFCD05864664
InChI Key FGJACYJASSSXNJ-UHFFFAOYSA-N
Synonym 1-boc-4-2-formylphenyl piperazine, tert-butyl 4-2-formylphenyl piperazine-1-carboxylate, 4-2-formylphenyl piperazine-1-carboxylic acid tert-butyl ester, tert-butyl 4-2-formylphenyl piperazinecarboxylate, 4-2-formyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, acmc-20a0zo, n-boc-4-2-formylphenyl-piperazine, 1-n-boc-4-2-formylphenyl piperazine, 1-boc-4-2-formyl-phenyl-piperazine
PubChem CID 6490990
IUPAC Name tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2C=O

Specifications

Melting Point 86°C to 90°C
Quantity 5 g
UN Number UN2811
Formula Weight 290.36
Percent Purity 97%
Chemical Name or Material 1-Boc-4-(2-formylphenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'