missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(2-methoxycarbonylphenyl)piperazine, 97%, Thermo Scientific™

Product Code. 15441518
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870703-74-5
Molecular Formula C17H24N2O4
Molecular Weight (g/mol) 320.39
MDL Number MFCD07369760
InChI Key JFZGCXORLWXLGH-UHFFFAOYSA-N
Synonym 1-boc-4-2-methoxycarbonylphenyl piperazine, tert-butyl 4-2-methoxycarbonyl phenyl piperazine-1-carboxylate, methyl 2-boc-piperazin-1-yl benzoate, 1-boc-4-2-methoxycarbonylphenyl pipera, 1-boc-4-2-methoxycarbonyl phenyl-piperazine
PubChem CID 16217924
IUPAC Name tert-butyl 4-(2-methoxycarbonylphenyl)piperazine-1-carboxylate
SMILES COC(=O)C1=CC=CC=C1N1CCN(CC1)C(=O)OC(C)(C)C

Specifications

Color Yellow to Orange
Flash Point >110°C (230°F)
Refractive Index 1.532
Quantity 1 g
UN Number UN2810
Formula Weight 320.39
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Boc-4-(2-methoxycarbonylphenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'