missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(4-aminophenyl)piperazine, 97%, Thermo Scientific Chemicals

Product Code. 15479748
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Boc-4-(4-aminophenyl)piperazine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-4-(4-aminophenyl)piperazine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170911-92-9
Molecular Formula C15H23N3O2
Molecular Weight (g/mol) 277.368
MDL Number MFCD04115065
InChI Key RXFHRKPNLPBDGE-UHFFFAOYSA-N
Synonym tert-butyl 4-4-aminophenyl piperazine-1-carboxylate, 4-4-aminophenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-piperazin-1-yl aniline, 1-boc-4-4-aminophenyl piperazine, 1-piperazinecarboxylic acid, 4-4-aminophenyl-, 1,1-dimethylethyl ester, 4-4-aminophenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-boc-4-4'-aminophenyl piperazine dihydrochloride, 1-boc-4-4-aminophenyl-piperazine, pubchem12198
PubChem CID 11011301
IUPAC Name tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)N

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 277.36
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(4-aminophenyl)piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'