Learn More
1-Boc-4-(4-aminophenyl)piperazine, 97%, Thermo Scientific Chemicals
POA - £242.00
Chemical Identifiers
CAS | 170911-92-9 |
---|---|
Molecular Formula | C15H23N3O2 |
Molecular Weight (g/mol) | 277.368 |
MDL Number | MFCD04115065 |
InChI Key | RXFHRKPNLPBDGE-UHFFFAOYSA-N |
Synonym | tert-butyl 4-4-aminophenyl piperazine-1-carboxylate, 4-4-aminophenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-piperazin-1-yl aniline, 1-boc-4-4-aminophenyl piperazine, 1-piperazinecarboxylic acid, 4-4-aminophenyl-, 1,1-dimethylethyl ester, 4-4-aminophenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-boc-4-4'-aminophenyl piperazine dihydrochloride, 1-boc-4-4-aminophenyl-piperazine, pubchem12198 |
PubChem CID | 11011301 |
IUPAC Name | tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15489748
|
Thermo Scientific Alfa Aesar
H63191.MD |
250 mg | |||||||
15469748
|
Thermo Scientific Alfa Aesar
H63191.03 |
1 g |
£66.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15479748
|
Thermo Scientific Alfa Aesar
H63191.06 |
5 g |
£242.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Boc-4-(4-aminophenyl)piperazine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Boc-4-(4-aminophenyl)piperazine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
170911-92-9 | |
277.368 | |
RXFHRKPNLPBDGE-UHFFFAOYSA-N | |
11011301 | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)N |
C15H23N3O2 | |
MFCD04115065 | |
tert-butyl 4-4-aminophenyl piperazine-1-carboxylate, 4-4-aminophenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-piperazin-1-yl aniline, 1-boc-4-4-aminophenyl piperazine, 1-piperazinecarboxylic acid, 4-4-aminophenyl-, 1,1-dimethylethyl ester, 4-4-aminophenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-boc-4-4'-aminophenyl piperazine dihydrochloride, 1-boc-4-4-aminophenyl-piperazine, pubchem12198 | |
tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate |
Specifications
170911-92-9 | |
MFCD04115065 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)N | |
277.368 | |
277.36 | |
Solid |
C15H23N3O2 | |
tert-butyl 4-4-aminophenyl piperazine-1-carboxylate, 4-4-aminophenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-piperazin-1-yl aniline, 1-boc-4-4-aminophenyl piperazine, 1-piperazinecarboxylic acid, 4-4-aminophenyl-, 1,1-dimethylethyl ester, 4-4-aminophenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-boc-4-4'-aminophenyl piperazine dihydrochloride, 1-boc-4-4-aminophenyl-piperazine, pubchem12198 | |
RXFHRKPNLPBDGE-UHFFFAOYSA-N | |
tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate | |
11011301 | |
97% | |
1-Boc-4-(4-aminophenyl)piperazine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only