Learn More
1-BOC-4-iodo-piperidine, 97%, Thermo Scientific Chemicals
£69.80 - £204.00
Chemical Identifiers
CAS | 301673-14-3 |
---|---|
Molecular Formula | C10H18INO2 |
Molecular Weight (g/mol) | 311.16 |
MDL Number | MFCD04115041 |
InChI Key | YFWQFKUQVJNPKP-UHFFFAOYSA-N |
Synonym | n-boc-4-iodopiperidine, 1-boc-4-iodopiperidine, n-boc-4-iodo-piperidine, 1-tert-butoxycarbonyl-4-iodopiperidine, tert-butyl 4-iodotetrahydro-1 2h-pyridinecarboxylate, 1-n-boc-4-iodopiperidine, 4-iodo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-iodopiperidinecarboxylate, tert-butyl 4-iodo-1-piperidinecarboxylate |
PubChem CID | 10892302 |
IUPAC Name | tert-butyl 4-iodopiperidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCC(CC1)I |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10725462
|
Thermo Scientific Acros
437270010 |
1 g | Glass bottle |
£69.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10735462
|
Thermo Scientific Acros
437270050 |
5 g | Glass bottle |
£204.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
301673-14-3 | |
311.16 | |
YFWQFKUQVJNPKP-UHFFFAOYSA-N | |
10892302 | |
CC(C)(C)OC(=O)N1CCC(CC1)I |
C10H18INO2 | |
MFCD04115041 | |
n-boc-4-iodopiperidine, 1-boc-4-iodopiperidine, n-boc-4-iodo-piperidine, 1-tert-butoxycarbonyl-4-iodopiperidine, tert-butyl 4-iodotetrahydro-1 2h-pyridinecarboxylate, 1-n-boc-4-iodopiperidine, 4-iodo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-iodopiperidinecarboxylate, tert-butyl 4-iodo-1-piperidinecarboxylate | |
tert-butyl 4-iodopiperidine-1-carboxylate |
Specifications
301673-14-3 | |
100.0 | |
Brown to White | |
96% min. (GC) | |
MFCD04115041 | |
YFWQFKUQVJNPKP-UHFFFAOYSA-N | |
tert-butyl 4-iodopiperidine-1-carboxylate | |
10892302 | |
97% | |
1-BOC-4-iodo-piperidine |
96.0 | |
44°C to 49°C | |
Authentic | |
C10H18INO2 | |
n-boc-4-iodopiperidine, 1-boc-4-iodopiperidine, n-boc-4-iodo-piperidine, 1-tert-butoxycarbonyl-4-iodopiperidine, tert-butyl 4-iodotetrahydro-1 2h-pyridinecarboxylate, 1-n-boc-4-iodopiperidine, 4-iodo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-iodopiperidinecarboxylate, tert-butyl 4-iodo-1-piperidinecarboxylate | |
CC(C)(C)OC(=O)N1CCC(CC1)I | |
311.16 | |
311.16 | |
Crystalline Powder or Low Melting Solid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
- 1-BOC-4-iodo-piperidine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362-Take off contaminated clothing.
RUO – Research Use Only