missing translation for 'onlineSavingsMsg'
Learn More

1-(Boc-amino)cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 15448588
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88950-64-5
Molecular Formula C9H15NO4
Molecular Weight (g/mol) 201.22
MDL Number MFCD00083257
InChI Key DSKCOVBHIFAJRI-UHFFFAOYSA-N
Synonym 1-boc-amino cyclopropanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropanecarboxylic acid, n-boc-1-aminocyclopropanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopropanecarboxylic acid, boc-acpc-oh, 1-tert-butoxy carbonyl amino cyclopropane-1-carboxylic acid, 1-boc amino cyclopanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 1-1,1-dimethylethoxy carbonyl amino
PubChem CID 688396
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopropane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CC1)C(O)=O

Specifications

Melting Point 118°C (decomposition)
Beilstein 4430628
Quantity 1 g
Formula Weight 201.22
Percent Purity 98%
Chemical Name or Material 1-(Boc-amino)cyclopropanecarboxylic acid
compliance-icons
Product Identifier
  • 1-(Boc-amino)cyclopropanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 1-(Boc-amino)cyclopropanecarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(Boc-amino)cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.