missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-indole, 97%, Thermo Scientific Chemicals

Product Code. 11358546
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75400-67-8
Molecular Formula C13H15NO2
Molecular Weight (g/mol) 217.27
MDL Number MFCD02093939
InChI Key OWPIFQXNMLDXKW-UHFFFAOYSA-N
Synonym 1-boc-indole, tert-butyl 1h-indole-1-carboxylate, n-boc-indole, tert-butyl 1-indolecarboxylate, 1h-indole-1-carboxylic acid, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl indole, n-boc indole, pubchem7228, acmc-1bjgd, 1-tert-butoxycarbonylindole
PubChem CID 3532980
SMILES CC(C)(C)OC(=O)N1C=CC2=CC=CC=C12

Specifications

Density 1.07
Boiling Point 113°C to 114°C (0.2 mmHg)
Refractive Index 1.543
Quantity 1 g
Beilstein 4674239
IUPAC Name tert-butyl 1H-indole-1-carboxylate
Formula Weight 217.27
Percent Purity 97%
Chemical Name or Material 1-Boc-indole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'