missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-N-Fmoc-D-tryptophan, 98%

Product Code. 15468738
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15468738

Brand: Thermo Scientific Alfa Aesar H62185.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 163619-04-3 | C31H30N2O6 | 526.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163619-04-3
Molecular Formula C31H30N2O6
Molecular Weight (g/mol) 526.59
MDL Number MFCD00153367
InChI Key ADOHASQZJSJZBT-AREMUKBSSA-N
Synonym fmoc-d-trp boc-oh, n, a-fmoc-n in-boc-d-tryptophan, fmoc-d-trp boc, ambotzfaa1339, pubchem10051, 1-boc-fmoc-d-tryptophan, 1-boc-n-fmoc-d-tryptophan, 2r-3-1-tert-butoxycarbonylindol-3-yl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, nalpha-fmoc-n in-boc-d-tryptophan, fmoc-d-trp boc-oh hplc
PubChem CID 16213150
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid
SMILES CC(C)(C)OC(=O)N1C=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12

Specifications

Quantity 5 g
Beilstein 7062970
Formula Weight 526.59
Percent Purity 98%
Chemical Name or Material 1-Boc-N-Fmoc-D-tryptophan

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.