Learn More
1-Bromo-2,2-dimethoxypropane, 98%, Thermo Scientific Chemicals
£26.73 - £342.71
Chemical Identifiers
CAS | 126-38-5 |
---|---|
Molecular Formula | C5H11BrO2 |
Molecular Weight (g/mol) | 183.045 |
MDL Number | MFCD00000208 |
InChI Key | SGTITUFGCGGICE-UHFFFAOYSA-N |
Synonym | propane,1-bromo-2,2-dimethoxy, propane, 1-bromo-2,2-dimethoxy, bromoacetone dimethyl acetal, bromo-2,2dimethoxypropane, bromo-2,2-dimethoxypropane, 1-bromo-2,2-dimethoxypropan, 1-bromo-2,2-dimethoxyprpopane, 1-bromo-2,2-dimethoxy propane, 1-bromo-2,2-dimethoxy-propane, bromoacetone dimethyl ketal |
PubChem CID | 67175 |
IUPAC Name | 1-bromo-2,2-dimethoxypropane |
SMILES | CC(CBr)(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449043
|
Thermo Scientific Alfa Aesar
A11139.09 |
10 g |
£26.73
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459043
|
Thermo Scientific Alfa Aesar
A11139.18 |
50 g |
£88.14
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11469043
|
Thermo Scientific Alfa Aesar
A11139.30 |
250 g |
£342.71
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
126-38-5 | |
183.045 | |
SGTITUFGCGGICE-UHFFFAOYSA-N | |
67175 | |
CC(CBr)(OC)OC |
C5H11BrO2 | |
MFCD00000208 | |
propane,1-bromo-2,2-dimethoxy, propane, 1-bromo-2,2-dimethoxy, bromoacetone dimethyl acetal, bromo-2,2dimethoxypropane, bromo-2,2-dimethoxypropane, 1-bromo-2,2-dimethoxypropan, 1-bromo-2,2-dimethoxyprpopane, 1-bromo-2,2-dimethoxy propane, 1-bromo-2,2-dimethoxy-propane, bromoacetone dimethyl ketal | |
1-bromo-2,2-dimethoxypropane |
Specifications
126-38-5 | |
83°C to 87°C (80 mmHg) | |
C5H11BrO2 | |
MFCD00000208 | |
2036095 | |
propane,1-bromo-2,2-dimethoxy, propane, 1-bromo-2,2-dimethoxy, bromoacetone dimethyl acetal, bromo-2,2dimethoxypropane, bromo-2,2-dimethoxypropane, 1-bromo-2,2-dimethoxypropan, 1-bromo-2,2-dimethoxyprpopane, 1-bromo-2,2-dimethoxy propane, 1-bromo-2,2-dimethoxy-propane, bromoacetone dimethyl ketal | |
CC(CBr)(OC)OC | |
183.045 | |
183.05 | |
1-Bromo-2,2-dimethoxypropane |
1.354 | |
40°C (104°F) | |
1.447 | |
UN3271 | |
Moisture Sensitive | |
SGTITUFGCGGICE-UHFFFAOYSA-N | |
1-bromo-2,2-dimethoxypropane | |
67175 | |
98% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
missing translation for 'einecsNumber' : 204-784-7
missing translation for 'tsca' : No
RUO – Research Use Only