Learn More
1-Bromo-2,4,5-trifluorobenzene, 98+%, Thermo Scientific™
£10.90 - £117.00
Chemical Identifiers
CAS | 327-52-6 |
---|---|
Molecular Formula | C6H2BrF3 |
Molecular Weight (g/mol) | 210.981 |
MDL Number | MFCD00000306 |
InChI Key | DVTULTINXNWGJY-UHFFFAOYSA-N |
Synonym | 2,4,5-trifluorobromobenzene, 2-bromo-1,4,5-trifluorobenzene, benzene, 1-bromo-2,4,5-trifluoro, 1-bromo-2,4,5-trifluoro-benzene, paragos 390205, 5-bromo-1,2,4-trifluorobenzene, fr bf df ee, pubchem2306, acmc-1cs6j, ksc224g3h |
PubChem CID | 67597 |
IUPAC Name | 1-bromo-2,4,5-trifluorobenzene |
SMILES | C1=C(C(=CC(=C1F)Br)F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications1-Bromo-2,4,5-trifluorobenzene is the intermediate of liquid crystals. The derivatives of this compound can be used as intermediates for the synthesis of qunolone antibacterial. It is employed in the synthesis of 3-ethenyl, 3-ethynyl, 3-aryl, and 3-cyclopropyl-2, 4, 5-trifluorobenzoic acids from 1-bromo-2, 4, 5-trifluorobenzene and 2, 4, 5-trifluoro-3-hydroxybenzoic acid.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
327-52-6 | |
210.981 | |
DVTULTINXNWGJY-UHFFFAOYSA-N | |
67597 | |
C1=C(C(=CC(=C1F)Br)F)F |
C6H2BrF3 | |
MFCD00000306 | |
2,4,5-trifluorobromobenzene, 2-bromo-1,4,5-trifluorobenzene, benzene, 1-bromo-2,4,5-trifluoro, 1-bromo-2,4,5-trifluoro-benzene, paragos 390205, 5-bromo-1,2,4-trifluorobenzene, fr bf df ee, pubchem2306, acmc-1cs6j, ksc224g3h | |
1-bromo-2,4,5-trifluorobenzene |
Specifications
327-52-6 | |
1.806 | |
55°C (131°F) | |
1.486 | |
UN1993 | |
2,4,5-trifluorobromobenzene, 2-bromo-1,4,5-trifluorobenzene, benzene, 1-bromo-2,4,5-trifluoro, 1-bromo-2,4,5-trifluoro-benzene, paragos 390205, 5-bromo-1,2,4-trifluorobenzene, fr bf df ee, pubchem2306, acmc-1cs6j, ksc224g3h | |
DVTULTINXNWGJY-UHFFFAOYSA-N | |
1-bromo-2,4,5-trifluorobenzene | |
67597 | |
≥98% |
-19°C | |
143°C to 144°C | |
C6H2BrF3 | |
MFCD00000306 | |
1636588 | |
Insoluble in water. | |
C1=C(C(=CC(=C1F)Br)F)F | |
210.981 | |
210.98 | |
1-Bromo-2,4,5-trifluorobenzene |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 206-318-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures