Learn More
1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific Chemicals
£41.70 - £537.00
Chemical Identifiers
CAS | 816-40-0 |
---|---|
Molecular Formula | C4H7BrO |
Molecular Weight (g/mol) | 151 |
MDL Number | MFCD00000207 |
InChI Key | CCXQVBSQUQCEEO-UHFFFAOYSA-N |
Synonym | 1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one |
PubChem CID | 13156 |
IUPAC Name | 1-bromobutan-2-one |
SMILES | CCC(=O)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10119612
|
Thermo Scientific Acros
222450010 |
1 g | Glass bottle |
£41.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10495993
|
Thermo Scientific Acros
222450050 |
5 g | Glass bottle |
£139.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10781725
|
Thermo Scientific Acros
222450250 |
25 g | Glass bottle |
£537.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
816-40-0 | |
151 | |
CCXQVBSQUQCEEO-UHFFFAOYSA-N | |
13156 | |
CCC(=O)CBr |
C4H7BrO | |
MFCD00000207 | |
1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one | |
1-bromobutan-2-one |
Specifications
816-40-0 | |
100.0 | |
1.4790g/mL | |
68°C | |
88% min. (GC) | |
C2H5COCH2Br | |
MFCD00000207 | |
1.479 | |
CCXQVBSQUQCEEO-UHFFFAOYSA-N | |
1-bromobutan-2-one | |
151 | |
151 | |
Liquid |
88.0 | |
Colorless to Yellow | |
105°C (150.0 mmHg) | |
Authentic | |
C4H7BrO | |
1.4650 to 1.469 | |
01,67 | |
1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one | |
CCC(=O)CBr | |
approx. 0.5% CaCO3 | |
13156 | |
90% | |
1-Bromo-2-butanone, Stabilized |
Safety and Handling
GHS H Statement
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-431-3
- 1-Bromo-2-butanone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only