missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-fluoro-4-nitrobenzene, 98%, Thermo Scientific Chemicals

£76.78 - £217.08

Chemical Identifiers

CAS 185331-69-5
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 220.00
MDL Number MFCD05865088
InChI Key AZYQMHQOHNGPRR-UHFFFAOYSA-N
Synonym 4-bromo-3-fluoronitrobenzene, 3-fluoro-4-bromonitrobenzene, benzene, 1-bromo-2-fluoro-4-nitro, pubchem1045, acmc-1byt6, ksc495e5r, 1-bromo-2-fluoro-4-nitro-benzene, 4-bromo-3-fluoronitrobenzene; 3-fluoro-4-bromonitrobenzene
PubChem CID 2756994
IUPAC Name 1-bromo-2-fluoro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(Br)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15438048
View Documents
Thermo Scientific Alfa Aesar
H61494.14
25 g
£76.78
25g
Please to purchase this item. Need a web account? Register with us today!
15448048
View Documents
Thermo Scientific Alfa Aesar
H61494.22
100 g
£217.08
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

185331-69-5
220.00
AZYQMHQOHNGPRR-UHFFFAOYSA-N
2756994
[O-][N+](=O)C1=CC(F)=C(Br)C=C1
C6H3BrFNO2
MFCD05865088
4-bromo-3-fluoronitrobenzene, 3-fluoro-4-bromonitrobenzene, benzene, 1-bromo-2-fluoro-4-nitro, pubchem1045, acmc-1byt6, ksc495e5r, 1-bromo-2-fluoro-4-nitro-benzene, 4-bromo-3-fluoronitrobenzene; 3-fluoro-4-bromonitrobenzene
1-bromo-2-fluoro-4-nitrobenzene

Specifications

185331-69-5
C6H3BrFNO2
4-bromo-3-fluoronitrobenzene, 3-fluoro-4-bromonitrobenzene, benzene, 1-bromo-2-fluoro-4-nitro, pubchem1045, acmc-1byt6, ksc495e5r, 1-bromo-2-fluoro-4-nitro-benzene, 4-bromo-3-fluoronitrobenzene; 3-fluoro-4-bromonitrobenzene
[O-][N+](=O)C1=CC(F)=C(Br)C=C1
220.00
220
Solid
88°C to 94°C
MFCD05865088
AZYQMHQOHNGPRR-UHFFFAOYSA-N
1-bromo-2-fluoro-4-nitrobenzene
2756994
98%
1-Bromo-2-fluoro-4-nitrobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.