Learn More
1-Bromo-2-methylnaphthalene, tech. 90%, Thermo Scientific Chemicals
£20.10 - £57.50
Chemical Identifiers
CAS | 2586-62-1 |
---|---|
Molecular Formula | C11H9Br |
Molecular Weight (g/mol) | 221.10 |
MDL Number | MFCD00003871 |
InChI Key | CMIMBQIBIZZZHQ-UHFFFAOYSA-N |
Synonym | naphthalene, 1-bromo-2-methyl, 2-methyl-1-bromonaphthalene, .beta.-methyl-.alpha.-bromonaphthalene, zlchem 421, 1-bromo-2-methylnaphtalene, acmc-1cq62, 1-bromo-2-methyl-naphthalene, 1-bromo-2-methyl naphthalene, 1-bromanyl-2-methyl-naphthalene, 1-bromonaphthalen-2-yl methyl |
PubChem CID | 75754 |
IUPAC Name | 1-bromo-2-methylnaphthalene |
SMILES | CC1=CC=C2C=CC=CC2=C1Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11331827
|
Thermo Scientific Alfa Aesar
L05093.09 |
10 g |
£20.10
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11341827
|
Thermo Scientific Alfa Aesar
L05093.18 |
50 g |
£57.50
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromo-2-methylnaphthalene is used as test compound in determination of halogenated hydrocarbons at trace levels by supercritical fluid chromatography-microwave-induced plasma mass spectrometry. Dynamic phosphorescence quenching of 1-bromo-2-methylnaphthalene without deoxygenation has been used for selective sensing of Cu(II) at ngm levels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-2-methylnaphthalene is used as test compound in determination of halogenated hydrocarbons at trace levels by supercritical fluid chromatography-microwave-induced plasma mass spectrometry. Dynamic phosphorescence quenching of 1-bromo-2-methylnaphthalene without deoxygenation has been used for selective sensing of Cu(II) at ngm levels.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
Chemical Identifiers
2586-62-1 | |
221.10 | |
CMIMBQIBIZZZHQ-UHFFFAOYSA-N | |
75754 | |
CC1=CC=C2C=CC=CC2=C1Br |
C11H9Br | |
MFCD00003871 | |
naphthalene, 1-bromo-2-methyl, 2-methyl-1-bromonaphthalene, .beta.-methyl-.alpha.-bromonaphthalene, zlchem 421, 1-bromo-2-methylnaphtalene, acmc-1cq62, 1-bromo-2-methyl-naphthalene, 1-bromo-2-methyl naphthalene, 1-bromanyl-2-methyl-naphthalene, 1-bromonaphthalen-2-yl methyl | |
1-bromo-2-methylnaphthalene |
Specifications
2586-62-1 | |
295°C to 296°C | |
C11H9Br | |
MFCD00003871 | |
naphthalene, 1-bromo-2-methyl, 2-methyl-1-bromonaphthalene, .beta.-methyl-.alpha.-bromonaphthalene, zlchem 421, 1-bromo-2-methylnaphtalene, acmc-1cq62, 1-bromo-2-methyl-naphthalene, 1-bromo-2-methyl naphthalene, 1-bromanyl-2-methyl-naphthalene, 1-bromonaphthalen-2-yl methyl | |
CMIMBQIBIZZZHQ-UHFFFAOYSA-N | |
1-bromo-2-methylnaphthalene | |
75754 | |
90% | |
1-Bromo-2-methylnaphthalene |
1.428 | |
>110°C (230°F) | |
1.649 | |
2042531 | |
Not miscible in water. | |
CC1=CC=C2C=CC=CC2=C1Br | |
221.10 | |
221.1 | |
Technical |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 219-966-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only