missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methylpropane, 98+%

Product Code. 10062110 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10062110

Brand: Thermo Scientific Acros 168835000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-77-3 | C4H9Br | 137.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-77-3
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
InChI Key HLVFKOKELQSXIQ-UHFFFAOYSA-N
Synonym isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal
PubChem CID 6555
IUPAC Name 1-bromo-2-methylpropane
SMILES CC(C)CBr

Specifications

Melting Point -119.0°C
Color Colorless
Density 1.2600g/mL
Boiling Point 90.0°C to 92.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2Br
Refractive Index 1.4346 to 1.4366
Quantity 500 mL
Beilstein 01,126
Merck Index 15,518
Specific Gravity 1.26
Solubility Information Solubility in water: 0.51g/L (18°C). Other solubilities: soluble in most common organic solvents
Formula Weight 137.02
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-methylpropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-2-methylpropane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.