missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methylpropane, 98+%

Product Code. 11486696
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11486696

Brand: Thermo Scientific Alfa Aesar B23854.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-77-3 | C4H9Br | 137.02 g/mol

1-Bromo-2-methylpropane is used as a synthetic reagent for the preparation of 1-ethyl-1,3-isobutylimidazolium bromide by reaction with 1-ethylimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methylpropane is used as a synthetic reagent for the preparation of 1-ethyl-1,3-isobutylimidazolium bromide by reaction with 1-ethylimidazole.

Solubility
Slightly soluble in water. Miscible with alcohol, ether.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-77-3
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000217
InChI Key HLVFKOKELQSXIQ-UHFFFAOYSA-N
Synonym isobutyl bromide, iso-butyl bromide, propane, 1-bromo-2-methyl, bromoisobutane, i-butyl bromide, 1-bromo-2-methyl-propane, unii-5oec0bw987, ccris 349, 1-bromo-2-methyl propane, bromoacetaldehydedimethylacetal
PubChem CID 6555
IUPAC Name 1-bromo-2-methylpropane
SMILES CC(C)CBr

Specifications

Melting Point -118°C
Density 1.255
Boiling Point 90°C to 92°C
Flash Point 18°C (64°F)
Refractive Index 1.435
Quantity 1000 g
UN Number UN2342
Beilstein 1730915
Merck Index 14,5135
Solubility Information Slightly soluble in water; Miscible with alcohol,ether.
Formula Weight 137.02
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-2-methylpropane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.