missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-nitrobenzene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
1-Bromo-2-nitrobenzene is used in organic synthesis. It also can react with 2-methylamino-benzoic acid to get N-methyl-N-(2-nitro-phenyl)-anthranilic acid. And it is also used in the preparation of 4-methoxy-2?-nitrodiphenyl ether, 1-methoxy-3,5-bis-(2-nitro-phenoxy)benzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-2-nitrobenzene is used in organic synthesis. It also can react with 2-methylamino-benzoic acid to get N-methyl-N-(2-nitro-phenyl)-anthranilic acid. And it is also used in the preparation of 4-methoxy-2′-nitrodiphenyl ether, 1-methoxy-3,5-bis-(2-nitro-phenoxy)benzene.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
1-Bromo-2-nitrobenzene is used in organic synthesis. It also can react with 2-methylamino-benzoic acid to get N-methyl-N-(2-nitro-phenyl)-anthranilic acid. And it is also used in the preparation of 4-methoxy-2′-nitrodiphenyl ether, 1-methoxy-3,5-bis-(2-nitro-phenoxy)benzene.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 40°C to 44°C |
Boiling Point | 261°C |
Flash Point | 112°C (233°F) |
Quantity | 250 g |
UN Number | UN3459 |
Beilstein | 2044109 |
Solubility Information | Insoluble in water. |
Formula Weight | 202.01 |
Percent Purity | 99% |
Chemical Name or Material | 1-Bromo-2-nitrobenzene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'