missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-pentyne, 97%, Thermo Scientific Chemicals

Product Code. 11914501
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16400-32-1
Molecular Formula C5H7Br
Molecular Weight (g/mol) 147.02
MDL Number MFCD00236363
InChI Key VDHGRVFJBGRHMD-UHFFFAOYSA-N
Synonym 1-bromo-2-pentyne, 2-pentynyl bromide, 2-pentyne, 1-bromo, 1-bromo-pent-2-yne, 1-bromo-2-pentine, 2-pentyn-1-yl bromide, acmc-1c6g2
PubChem CID 85399
IUPAC Name 1-bromopent-2-yne
SMILES CCC#CCBr

Specifications

Color Yellow
Boiling Point 40°C to 41°C (20.0 mmHg)
Flash Point 36°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.508
Quantity 1 g
Formula Weight 147.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-pentyne
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-2-pentyne
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'