missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-pentyne, 97%, Thermo Scientific Chemicals

£19.22 - £86.60

Chemical Identifiers

CAS 16400-32-1
Molecular Formula C5H7Br
Molecular Weight (g/mol) 147.02
MDL Number MFCD00236363
InChI Key VDHGRVFJBGRHMD-UHFFFAOYSA-N
Synonym 1-bromo-2-pentyne, 2-pentynyl bromide, 2-pentyne, 1-bromo, 1-bromo-pent-2-yne, 1-bromo-2-pentine, 2-pentyn-1-yl bromide, acmc-1c6g2
PubChem CID 85399
IUPAC Name 1-bromopent-2-yne
SMILES CCC#CCBr
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11914501
View Documents
Thermo Scientific Acros
439570010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11924501
View Documents
Thermo Scientific Acros
439570100
10 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16400-32-1
147.02
VDHGRVFJBGRHMD-UHFFFAOYSA-N
85399
CCC#CCBr
C5H7Br
MFCD00236363
1-bromo-2-pentyne, 2-pentynyl bromide, 2-pentyne, 1-bromo, 1-bromo-pent-2-yne, 1-bromo-2-pentine, 2-pentyn-1-yl bromide, acmc-1c6g2
1-bromopent-2-yne

Specifications

16400-32-1
40°C to 41°C (20.0 mmHg)
96% min. (GC)
1.508
1-bromo-2-pentyne, 2-pentynyl bromide, 2-pentyne, 1-bromo, 1-bromo-pent-2-yne, 1-bromo-2-pentine, 2-pentyn-1-yl bromide, acmc-1c6g2
CCC#CCBr
147.02
147.01
Liquid
Yellow
36°C
C5H7Br
MFCD00236363
VDHGRVFJBGRHMD-UHFFFAOYSA-N
1-bromopent-2-yne
85399
97%
1-Bromo-2-pentyne
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair):

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 240-451-2

Product Identifier
  • 1-Bromo-2-pentyne

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only