missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,4,5-trichlorobenzene, 98%, Thermo Scientific Chemicals

Product Code. 15481749
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a multipurpose intermediate commonly used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a multipurpose intermediate commonly used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21928-51-8
Molecular Formula C6H2BrCl3
Molecular Weight (g/mol) 260.34
MDL Number MFCD00155009
InChI Key VZUMVBQMJFFYRM-UHFFFAOYSA-N
Synonym 3,4,5-trichlorobromobenzene, benzene, 5-bromo-1,2,3-trichloro, 1-bromo-3,4,5-trichlorobenzene, 1-bromo-3,4,5-trichloro-benzene, pubchem23062, acmc-209fpm, ksc497i6l, 1,2,3-trichloro-5-bromobenzene
PubChem CID 4124400
IUPAC Name 5-bromo-1,2,3-trichlorobenzene
SMILES ClC1=CC(Br)=CC(Cl)=C1Cl

Specifications

Color White
Quantity 100 g
Physical Form Powder
Beilstein 2089937
Solubility Information Slightly soluble in water.
Formula Weight 260.34
Percent Purity 98%
Chemical Name or Material 1-Bromo-3,4,5-trichlorobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'