missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -59.0°C |
Density | 1.5900g/mL |
Boiling Point | 142.0°C to 145.0°C |
Flash Point | 81°C |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | Cl(CH2)3Br |
Refractive Index | 1.4850 to 1.4870 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- 1-Bromo-3-chloropropane
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Germ cell mutagenicity Category 2
- Specific target organ toxicity Category 3
- H341-Suspected of causing genetic defects.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'