missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-(difluoromethoxy)benzene, 97+%, Thermo Scientific™

Product Code. 11486856
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 262587-05-3
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.017
MDL Number MFCD00236233
InChI Key NWBDGKNKAFGQQL-UHFFFAOYSA-N
Synonym 1-bromo-3-difluoromethoxy benzene, 3-difluoromethoxy bromobenzene, benzene, 1-bromo-3-difluoromethoxy, 1-difluoromethoxy-3-bromobenzene, pubchem18537, acmc-1crzp, 3-difluoromethoxybromobenzene, ksc493c1b, 3-bromo-1-difluoromethoxy benzene, 3-difluoromethyl bromobenzene
PubChem CID 2737006
IUPAC Name 1-bromo-3-(difluoromethoxy)benzene
SMILES C1=CC(=CC(=C1)Br)OC(F)F

Specifications

Quantity 1 g
Formula Weight 223.02
Percent Purity ≥97%
Chemical Name or Material 1-Bromo-3-(difluoromethoxy)benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'