missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-fluorobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11399206
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-06-9
Molecular Formula C6H4BrF
MDL Number MFCD00000326
InChI Key QDFKKJYEIFBEFC-UHFFFAOYSA-N
Synonym 3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo
PubChem CID 14082
IUPAC Name 1-bromo-3-fluorobenzene

Specifications

Melting Point -8°C
Density 1.594
Boiling Point 148°C to 150°C
Flash Point 46°C
Odor Aromatic
Refractive Index 1.5275
Quantity 10 g
UN Number UN1993
Beilstein 1853969
SMILES FC1=CC=CC(Br)=C1
Molecular Weight (g/mol) 175.00
Formula Weight 175.01
Percent Purity 99%
Chemical Name or Material 1-Bromo-3-fluorobenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.