missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3-isopropylbenzene, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Difficult to mix in water.
Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Difficult to mix in water.
Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.285 |
Boiling Point | 210°C |
Molecular Formula | C9H11Br |
Refractive Index | 1.534 |
Quantity | 1 g |
Beilstein | 1858092 |
Synonym | 1-bromo-3-isopropylbenzene, 3-isopropylbromobenzene, 3-bromocumene, 3-bromoisopropylbenzene, benzene, 1-bromo-3-1-methylethyl, 1-bromo-3-isopropyl-benzene, 1-bromo-3-1-methylethyl benzene, m-bromocumene, m-isopropylbromobenzene, m-bromo-isopropyl-benzene |
Solubility Information | Difficult to mix in water. |
InChI Key | GBSGGFCCQZUXNB-UHFFFAOYSA-N |
SMILES | CC(C)C1=CC(=CC=C1)Br |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'