missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3-methyl-2-butene, 90+%, stab. with silver, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Beschreibung
1-Bromo-3-methyl-2-butene is considered as a source of 3,3-dimethylallyl group and used in the preparation of natural products. It is also employed in the synthesis of base-modified pyrimidine nucleosides. Further, it is used in the preparation of (±)-eldanolide, 1-(3,3-dimethylallyl)-L-tryptophan and (±)-fumagillin, an antibiotic isolated from Aspergillus fumigattus.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-3-methyl-2-butene is considered as a source of 3,3-dimethylallyl group and used in the preparation of natural products. It is also employed in the synthesis of base-modified pyrimidine nucleosides. Further, it is used in the preparation of (±)-eldanolide, 1-(3,3-dimethylallyl)-L-tryptophan and (±)-fumagillin, an antibiotic isolated from Aspergillus fumigattus.
Solubility
Immiscible with water
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases
1-Bromo-3-methyl-2-butene is considered as a source of 3,3-dimethylallyl group and used in the preparation of natural products. It is also employed in the synthesis of base-modified pyrimidine nucleosides. Further, it is used in the preparation of (±)-eldanolide, 1-(3,3-dimethylallyl)-L-tryptophan and (±)-fumagillin, an antibiotic isolated from Aspergillus fumigattus.
Solubility
Immiscible with water
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases
Spezifikation
Spezifikation
Density | 1.281 |
Boiling Point | 135°C |
Flash Point | 32°C (90°F) |
Odor | Pungent |
Refractive Index | 1.4895 |
Quantity | 50 g |
UN Number | UN2920 |
Beilstein | 1420778 |
Sensitivity | Light sensitive |
Solubility Information | Immiscible with water |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'