Learn More
1-Bromo-3-methyl-2-butene, 96%, Thermo Scientific Chemicals
£33.37 - £107.54
Chemical Identifiers
CAS | 870-63-3 |
---|---|
Molecular Formula | C5H9Br |
Molecular Weight (g/mol) | 149.04 |
MDL Number | MFCD00000242 |
InChI Key | LOYZVRIHVZEDMW-UHFFFAOYSA-N |
Synonym | 1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide |
PubChem CID | 70092 |
IUPAC Name | 1-bromo-3-methylbut-2-ene |
SMILES | CC(=CCBr)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10105850
|
Thermo Scientific Acros
307410100 |
10 g |
£33.37
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15254704
|
Thermo Scientific Acros
307410500 |
50 g |
£107.54
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
870-63-3 | |
149.04 | |
LOYZVRIHVZEDMW-UHFFFAOYSA-N | |
70092 | |
CC(=CCBr)C |
C5H9Br | |
MFCD00000242 | |
1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide | |
1-bromo-3-methylbut-2-ene |
Specifications
870-63-3 | |
100.0 | |
1.2700g/mL | |
32°C | |
95% min. (ex Br) (Argentometry) | |
C5H9Br | |
1.4890 to 1.4910 | |
1.27 | |
LOYZVRIHVZEDMW-UHFFFAOYSA-N | |
1-bromo-3-methylbut-2-ene | |
149.04 | |
149.04 | |
Liquid |
95.0 | |
Brown or Yellow | |
82.0°C to 83.0°C (150.0 mmHg) | |
Authentic | |
Glass bottle | |
BrCH2CH=C(CH3)2 | |
MFCD00000242 | |
1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide | |
CC(=CCBr)C | |
silver wool | |
70092 | |
96% | |
1-Bromo-3-methyl-2-butene, 96% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-799-5
missing translation for 'tsca' : TSCA
- 1-Bromo-3-methyl-2-butene
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only